Hamilton SoftGrip™ Fixed-Volume Pipettor, Brick Red, 300 µL
Representative image only.
Hamilton SoftGrip™ Fixed-Volume Pipettor, Brick Red, 300 µL
Hamilton | Mfr: 55019-15| Item: 2570437
Contoured,ergonomical design provides comfortable, fatigue-free operation
₹60,100.00INR / Each
for an additional
₹14,500.00INR / Each
for an additional
₹25,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹60,100.00INR / Each
Buy It Together
Current Item
Hamilton SoftGrip™ Fixed-Volume Pipettor, Brick Red, 300 µL
Hamilton SoftGrip™ Fixed-Volume Pipettor, Brick Red, 300 µL
2570437
₹60,100.00INR / Each
+ =
Total Price 74600.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Color coding allows easy volume identification
  • Autoclave entire pipettor for total decontamination and sterilization
  • 1 year warranty
More About this Item
SoftGrip™ pipettors assure you of quality, comfort and precision manual pipetting. The slim ejection sleeve is made from a high strength, chemically resistant polymer. The slim design allows the pipette to reach to the bottom of narrow tubes without the need for extended length tips. SoftGrip pipettes have a universal calibration key that reduces the need for costly calibration services. Calibration requires only minutes to perform and eliminates the downtime and cost of alternative calibration methods. The SoftGrip tip nozzle ensures a proper fit with universal pipette tips. Unique color coding and labeling clearly indicates the volume of the pipettes.
1 Year Warranty
Specifications
  • SterileNo
  • AutoclavableYes
  • Max Volume (µL)300
  • Accuracy (%)±0.40%
  • Reproducibility0.0018
  • Number Of Channels1
  • Tip EjectorYes
  • Color CodeBrick Red
  • DescriptionFixed-Volume Pipettor, Brick Red, 300 µL