Hamilton® Gastight® Syringes with Luer Tip
Gas- and liquid-tight up to 200 psi!
  • Syringes handle corrosive gases and liquids, radioactive materials, and sterile solutions—ideal for gas and liquid chromatography
  • Sample loss is negligible at pressures up to 200 psi
These essentially inert syringes have barrels and plungers that are interchangeable within each syringe size, allowing field repair. Syringes with Luer Tip Connections feature a ground glass, luer taper. Autoclavable if plunger is removed. Needles are not included, order separately.
11,600.00 - ₹26,600.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Syringe Capacity (µL) Description
Availability Pricing
Hamilton Gastight® Syringe with Luer Tip, 10 µL, 1000 psig 0793920 Mfr # 80001
COMPARE
Borosilicate glass, PTFE 10 Syringe with Luer Tip, 10 µL, 1000 psig
₹11,600.00INR / Each
Hamilton Gastight® Syringe with Luer Tip, 25 µL, 1000 psig 0793921 Mfr # 80201
COMPARE
Borosilicate glass, PTFE 25 Syringe with Luer Tip, 25 µL, 1000 psig
₹12,500.00INR / Each
Hamilton Gastight® Syringe with Luer Tip, 50 µL, 1000 psig 0793922 Mfr # 80901
COMPARE
Borosilicate glass, PTFE 50 Syringe with Luer Tip, 50 µL, 1000 psig
₹12,500.00INR / Each
Hamilton Gastight® Syringe with Luer Tip, 100 µL, 1000 psig 0793923 Mfr # 81001
COMPARE
Borosilicate glass, PTFE 100 Syringe with Luer Tip, 100 µL, 1000 psig
₹12,500.00INR / Each
Hamilton Gastight® Syringe with Luer Tip, 250 µL, 500 psig 0793924 Mfr # 81101
COMPARE
Borosilicate glass, PTFE 250 Syringe with Luer Tip, 250 µL, 500 psig
₹13,200.00INR / Each
Hamilton Gastight® Syringe with Luer Tip, 500 µL, 700 psig 0793925 Mfr # 81201
COMPARE
Borosilicate glass, PTFE 500 Syringe with Luer Tip, 500 µL, 700 psig
₹14,800.00INR / Each
Hamilton Gastight® Syringe with Luer Tip, 1 mL, 200 psig 0793970 Mfr # 81301
COMPARE
Borosilicate glass, PTFE 1000 Syringe with Luer Tip, 1 mL, 200 psig
₹17,500.00INR / Each
Hamilton Gastight® Syringe with Luer Tip, 2.5 mL, 200 psig 0793972 Mfr # 81401
COMPARE
Borosilicate glass, PTFE 2500 Syringe with Luer Tip, 2.5 mL, 200 psig
₹18,500.00INR / Each
Hamilton Gastight® Syringe with Luer Tip, 5 mL, 200 psig 0793973 Mfr # 81501
COMPARE
Borosilicate glass, PTFE 5000 Syringe with Luer Tip, 5 mL, 200 psig
₹22,300.00INR / Each
Hamilton Gastight® Syringe with Luer Tip, 10 mL, 200 psig 0793974 Mfr # 81601
COMPARE
Borosilicate glass, PTFE, SST 10000 Syringe with Luer Tip, 10 mL, 200 psig
₹26,600.00INR / Each
Showing 1- 10 of 10