Hamilton® Chaney Adapter for Syringes
Assures repetitive and identical syringe plunger location
  • Eliminate subjective errors when setting a syringe plunger at a specific volume
Hamilton® Chaney Adapter ensures reporducibility of better than ±1% use when injections of identical volumes are required. A small set screw on one of the guide arms can be adjusted to provide a consistent fill volume for every dispense. This accessory also adds stability to the plunger minimizing the risk of a bent or broken plunger shaft.
19,000.00 - ₹22,900.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compatible With Syringe Type Description
Availability Pricing
Hamilton Chaney Adapter for 10 µL Syringes WW-07938-80 Mfr # 14700
COMPARE
10 µL syringes Chaney Adapter Chaney Adapter for 10 µL Syringes
₹22,900.00INR / Each
Hamilton Chaney Adapter for 25-500 µL Syringes WW-07938-81 Mfr # 14725
COMPARE
25 to 500 µL syringes Chaney Adapter Chaney Adapter for 25-500 µL Syringes
₹19,000.00INR / Each