Hamilton Chaney Adapter for 25-500 µL Syringes
Representative image only.
Hamilton Chaney Adapter for 25-500 µL Syringes
Hamilton | Mfr: 14725| Item: 0793881
Assures repetitive and identical syringe plunger location
₹33,500.00INR / Each
₹33,500.00INR / Each
Product Description
Key Features
  • Eliminate subjective errors when setting a syringe plunger at a specific volume
  • 18 month warranty
More About this Item
Hamilton® Chaney Adapter ensures reporducibility of better than ±1% use when injections of identical volumes are required. A small set screw on one of the guide arms can be adjusted to provide a consistent fill volume for every dispense. This accessory also adds stability to the plunger minimizing the risk of a bent or broken plunger shaft.
18 Month Warranty
Specifications
  • Syringe TypeChaney Adapter
  • Compatible With25 to 500 µL syringes
  • DescriptionChaney Adapter for 25-500 µL Syringes
  • Warranty18 months