Hach Water Quality Test Strips
Reduce risk by using as a pre-test validation tool
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
132,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Reagent Platform Test Tests Per Kit
Availability Pricing
Hach ARSENIC STRIPS Arsenic Low Range Test Strips, 100 (Range 0 to 500 ppb) 0554608 Mfr # 2800000
COMPARE
Strips Arsenic, low range 100
₹132,000.00INR / Each