Hach ARSENIC STRIPS Arsenic Low Range Test Strips, 100 (Range 0 to 500 ppb)
Representative image only.
Hach ARSENIC STRIPS Arsenic Low Range Test Strips, 100 (Range 0 to 500 ppb)
Hach | Mfr: 2800000| Item: 0554608
Reduce risk by using as a pre-test validation tool
₹132,000.00INR / Each
₹132,000.00INR / Each
Buy It Together
Current Item
Hach ARSENIC STRIPS Arsenic Low Range Test Strips, 100 (Range 0 to 500 ppb)
Hach ARSENIC STRIPS Arsenic Low Range Test Strips, 100 (Range 0 to 500 ppb)
0554608
₹132,000.00INR / Each
+ + +
Environmental Express Oakton pH Indicator Strips, 0 to 14, 100/pk
3585002
₹5,300.00INR / Pkg of 100
+ =
Total Price 160600.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Includes
test strips and color key for reading results.
Key Features
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
More About this Item
Test for arsenic from 0 to 500 ppb. 100 test strips included.
ISO9001 Certified Supplier
Specifications
  • TestArsenic, low range
  • Measurement Points0, 10, 30, 50, 70, 300, 500 ppb
  • Reagent PlatformStrips
  • Tests Per Kit100
  • DescriptionArsenic Low Range Test Strips, 100 (Range 0 to 500 ppb)