Hach HQ1130 Series Portable Dissolved Oxygen Meters
A robust and intuitive dissolved oxygen meter
  • Intellical™ smart probes have “plug-and-play” automatic parameter recognition
  • Easy step-by-step illustrated calibration and troubleshooting procedures reduce errors
  • Store up to 5,000 data points
  • Option to select from 27 languages in setup mode
  • IP67 rating with battery compartment installed
  • GLP-compliant features include time, date, sample ID, operator ID, and calibration
Hach HQ portable meters offer a versatile range of measurements, including dissolved oxygen (DO) and temperature. The Intellical™ probes are automatically recognized by the HQ meter and will retain calibration history and method settings to minimize errors and setup time. The proven technology delivers superior accuracy and response time, even when measuring challenging samples or in the field.
The illustrated screen displays step-by-step calibration and troubleshooting procedures on the 536 x 335 Mono-TFT display. Whether you are in the lab or in the field, transfer your measurements in just a few clicks via USB or optional Bluetooth. The optional Hach Communication Dongle (HCD) connects the meter to Claros™ data management software for additional data validation.
209,000.00 - ₹597,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Hach HQ1130  Portable Dedicated Dissolved Oxygen Meter WW-59005-05 Mfr # LEV015.53.1130A
COMPARE
Portable Dedicated Dissolved Oxygen Meter
₹209,000.00INR / Each
Hach HQ1130  Portable Dedicated Dissolved Oxygen Meter with DO Electrode, 1 m Cable WW-59005-06 Mfr # LEV015.53.11301
COMPARE
Portable Dedicated Dissolved Oxygen Meter with DO Electrode, 1 m Cable
₹551,000.00INR / Each
Hach HQ1130  Portable Dedicated Dissolved Oxygen Meter with DO Electrode, 5 m Rugged Cable WW-59005-07 Mfr # LEV015.53.11302
COMPARE
Portable Dedicated Dissolved Oxygen Meter with DO Electrode, 5 m Rugged Cable
₹597,000.00INR / Each