Hach HQ1130 Portable Dedicated Dissolved Oxygen Meter with DO Electrode, 5 m Rugged Cable

Representative image only.

Hach HQ1130 Portable Dedicated Dissolved Oxygen Meter with DO Electrode, 5 m Rugged Cable

Hach | Mfr: LEV015.53.11302| Item: 5900507

A robust and intuitive dissolved oxygen meter

₹836,000.00INR / Each

₹836,000.00INR / Each

Buy It Together

Current Item

Hach HQ1130  Portable Dedicated Dissolved Oxygen Meter with DO Electrode, 5 m Rugged Cable
Hach HQ1130 Portable Dedicated Dissolved Oxygen Meter with DO Electrode, 5 m Rugged Cable
5900507

₹836,000.00INR / Each
+
Hach LDO10101 IntelliCAL LDO Lab Probe, 1 m cable
5900457

₹350,000.00INR / Each
+ + =
Total Price 1666000.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Meter, LDO10105 DO electrode (59004-59), rechargeable battery, USB power adapter, and USB cable.

Key Features

  • Intellical™ smart probes have “plug-and-play” automatic parameter recognition
  • Easy step-by-step illustrated calibration and troubleshooting procedures reduce errors
  • Store up to 5,000 data points
  • Option to select from 27 languages in setup mode
  • IP67 rating with battery compartment installed
  • GLP-compliant features include time, date, sample ID, operator ID, and calibration
  • 5 m rugged cable DO electrode
  • 1 year warranty

More About this Item

Hach HQ portable meters offer a versatile range of measurements, including dissolved oxygen (DO) and temperature. The Intellical™ probes are automatically recognized by the HQ meter and will retain calibration history and method settings to minimize errors and setup time. The proven technology delivers superior accuracy and response time, even when measuring challenging samples or in the field.

The illustrated screen displays step-by-step calibration and troubleshooting procedures on the 536 x 335 Mono-TFT display. Whether you are in the lab or in the field, transfer your measurements in just a few clicks via USB or optional Bluetooth. The optional Hach Communication Dongle (HCD) connects the meter to Claros™ data management software for additional data validation.

1 Year WarrantyConformite Europeenne
Specifications
  • Meter TypeDissolved oxygen
  • Display type536x336 Mono-TFT
  • Buffer RecognitionIUPAC, USA, DIN, Custom
  • CalibrationUp to 3 points
  • Min Dissolved Oxygen0.0 mg/L
  • Max Dissolved Oxygen20 mg/L
  • Dissolved Oxygen Resolution0.1 mg/L
  • Dissolved Oxygen Accuracy±0.1 mg/L from 0 to 8 mg/L±0.2 mg/L for greater than 8 mg/L
  • Min Temperature (° C)-10
  • Max Temperature (° C)110
  • Min Temperature (° F)14
  • Max Temperature (° F)230
  • Temperature Resolution 0.1°C
  • Temperature Accuracy±0.3 °C (±0.54 °F)
  • Temperature compensationATC
  • OutputUSB
  • Data Logging (points)Yes
  • Memory Size5,000 data points
  • WaterproofIP67
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Battery1 rechargeable lithium-ion (included)
  • Length (cm)22
  • Width (cm)9.7
  • Height (cm)6.3
  • Width (in)3 13/16
  • Length (in)8 21/32
  • Height (in)2 31/64
  • Number of Positions1
  • NIST Calibration IncludedNo
  • Probe Included5 m rugged cable DO electrode
  • DescriptionPortable Dedicated Dissolved Oxygen Meter with DO Electrode, 5 m Rugged Cable
  • Warranty1 year
Reviews
Q&A