Hach® DR6000™ UV-Vis Spectrophotometer
Simplify your water analysis with guided procedures and optional RFID technology
  • Features over 250 pre-programmed procedures for water analysis
  • Operates in the UV and visible spectrum
  • TNTplus tests offer RFID technology that will automatically update your calibration factors when the test box is near the instrument
  • The instrument takes 10 readings of one sample to reduce error and remove outliers
This spectrophotometer revolutionizes instrument calibration with optional RFID technology. The RFID model will recognize a TNTplus test when placed near the scanner, automatically comparing your individual calibration information to the factory calibration curve. This results in a more efficient, accurate instrument. On top of that, the unit has 250 preprogrammed methods ready to go. This makes running any common test parameter easy and painless. The units have flexible connectivity and come with one ethernet port and three USB ports. It is already set up to interface with the Hach WIMS™ system for data management and storage.
6,232,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Hach DR6000 UV/VIS Spectrophotometer without RFID Technology 9951560 Mfr # LPV441.99.00002
COMPARE
UV/VIS Spectrophotometer without RFID Technology
₹6,232,000.00INR / Each
Hach DR6000 UV/VIS Spectrophotometer with RFID Technology 9951561 Mfr # LPV441.99.00012
COMPARE
UV/VIS Spectrophotometer with RFID Technology
₹6,232,000.00INR / Each