Hach® DR3900™ Spectrophotometer
Simplify your water analysis with guided procedures and optional RFID technology
  • Feature over 220 preprogrammed procedures for water analysis
  • Options available for EPA compliant test parameters
  • TNTplus tests offer RFID technology that will automatically update your calibration factors when the test box is placed near the instrument
  • The instrument takes 10 readings of one sample to reduce error and remove outliers
These spectrophotometers revolutionize instrument calibration with optional RFID technology. The RFID model will recognize a TNTplus test when placed near the scanner, automatically comparing your individual calibration information to the factory-installed calibration curve. This results in a more efficient, accurate instrument. On top of that, the unit has 220 preprogrammed methods ready to go. This makes running any common test parameter easy and painless. The units have flexible connectivity and come with one ethernet port and three USB ports. It is already set up to interface with the Hach WIMS™ system for data management and storage.
3,103,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Hach DR3900 Benchtop Visible Spectrophotometer 9951500 Mfr # LPV440.99.00002
COMPARE
Benchtop Visible Spectrophotometer
₹3,103,000.00INR / Each
Hach DR3900 Benchtop Visible Spectrophotometer with RFID 9951502 Mfr # LPV440.99.00012
COMPARE
Benchtop Visible Spectrophotometer with RFID
₹3,103,000.00INR / Each