H-B Instrument Enviro-Safe FRIO-Temp Nonhazardous PFA-Coated Verification Bottle Thermometers, Standard Accuracy,- Celsius Scale
Enclosed bottle ensures accuracy
  • Enviro-Safe is certified as 100% non-toxic, non-hazardous, biodegradable, and “green”
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy included
Accurately monitor temperatures in freezers, refrigerators, incubators, ovens, and other environments where stability is required. All Thermometers are filled with Enviro-Safe liquid and supplied with a unbreakable plastic bottle filled with Enviro-Safe liquid or glass beads. The thermometer can be removed from the bottle for recalibration if needed. Thermometers are PFA coated for enhanced safety in case of breakage. Use the magnetic or hook-and-loop attachment strip to securely attach the bottle to object such as a metal wall or door. All Enviro-Safe™ glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, RoHS compliant, individual serial numbered and have a statement of accuracy.
Call For Price
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Overall Length (mm)
Availability Pricing
H-B Instrument B61004-0400 FRIO-Temp Blood Bank Verification Thermometer; -5 to 20C 0807752 Mfr # B610040400
COMPARE
20 -5 135
Call For Price
H-B Instrument B61004-0300 FRIO-Temp Freezer Verification Thermometer; -30 to 1C 0807753 Mfr # B610040300
COMPARE
1 -30 135
Call For Price
H-B Instrument B61004-0100 FRIO-Temp Refrigerator Verification Thermometer; -5 to 15C 0807754 Mfr # B610040100
COMPARE
15 -5 125
Call For Price
H-B Instrument B61004-0500 FRIO-Temp Incubator Verification Thermometer; 15 to 50C 0807756 Mfr # B610040500
COMPARE
50 15 135
Call For Price
H-B Instrument B61004-1200 FRIO-Temp Ultra Low Freezer Verification Thermometer; -130 to 80F 9026060 Mfr # B610041200
COMPARE
145
Call For Price
H-B Instrument B61004-0900 FRIO-Temp Freezer Verification Thermometer; -20 to 35F 9026061 Mfr # B610040900
COMPARE
135
Call For Price
H-B Instrument B61004-1000 FRIO-Temp Blood Bank Verification Thermometer; 20 to 70F 9026062 Mfr # B610041000
COMPARE
135
Call For Price
H-B Instrument B61004-1100 FRIO-Temp Incubator Verification Thermometer; 60 to 120F 9026063 Mfr # B610041100
COMPARE
135
Call For Price
H-B Instrument B61001-0100 FRIO-Temp Freezer Verification Thermometer; -25 to -5C 9030060 Mfr # B610010100
COMPARE
-5 -25 210
Call For Price
H-B Instrument B61001-0200 FRIO-Temp Refrigerator Verification Thermometer; -2 to 10C 9030061 Mfr # B610010200
COMPARE
10 -2 180
Call For Price
H-B Instrument B61001-0300 FRIO-Temp Incubator Verification Thermometer; 15 to 30C 9030062 Mfr # B610010300
COMPARE
30 15 180
Call For Price
H-B Instrument B61001-0400 FRIO-Temp Incubator Verification Thermometer; 25 to 45C 9030063 Mfr # B610010400
COMPARE
45 25 190
Call For Price
H-B Instrument B61001-0500 FRIO-Temp Oven Verification Thermometer; 95 to 115C 9030064 Mfr # B610010500
COMPARE
115 95 200
Call For Price
H-B Instrument B61002-0100 FRIO-Temp Ultra Low Freezer Verification Thermometer; -90 to 25C 9030065 Mfr # B610040000
COMPARE
25 -90 145
Call For Price
Showing 1- 14 of 14