H-B Instrument B61002-0100 FRIO-Temp Ultra Low Freezer Verification Thermometer; -90 to 25C

Representative image only.

H-B Instrument B61002-0100 FRIO-Temp Ultra Low Freezer Verification Thermometer; -90 to 25C

H-B Instrument | Mfr: B610040000| Item: 9030065

Enclosed bottle ensures accuracy

Call For Price

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Call For Price

Buy It Together

Current Item

H-B Instrument B61002-0100 FRIO-Temp Ultra Low Freezer Verification Thermometer; -90 to 25C
H-B Instrument B61002-0100 FRIO-Temp Ultra Low Freezer Verification Thermometer; -90 to 25C
9030065

Call For Price
+ =
Total Price 33100.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Enviro-Safe is certified as 100% non-toxic, non-hazardous, biodegradable, and “green”
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy included
  • 5 year warranty

More About this Item

Accurately monitor temperatures in freezers, refrigerators, incubators, ovens, and other environments where stability is required. All Thermometers are filled with Enviro-Safe liquid and supplied with a unbreakable plastic bottle filled with Enviro-Safe liquid or glass beads. The thermometer can be removed from the bottle for recalibration if needed. Thermometers are PFA coated for enhanced safety in case of breakage. Use the magnetic or hook-and-loop attachment strip to securely attach the bottle to object such as a metal wall or door. All Enviro-Safe™ glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, RoHS compliant, individual serial numbered and have a statement of accuracy.

5 Year Warranty
Specifications
  • Length (")5 45/64
  • Min Temperature (° C)-90
  • Max Temperature (° C)25
  • Accuracy±1.0°C; ±2.0°C between -1°C and -40°C; ±2.5°C below -40°C
  • Division (° C)1
  • Thermometer FillToluene
  • Bottle FillGlass Beads
  • Overall Length (InchShort)5.7
  • Overall Length (mm)145
  • Stem MaterialPFA-Coated White-Back Glass
  • DescriptionFRIO-Temp Ultra Low Freezer Verification Thermometer; -90 to 25C
  • Warranty5 years
Reviews
Q&A