H-B Instrument Enviro-Safe Double-Safe Nonhazardous PFA-Coated Safety Glass Thermometers, Fahrenheit-scale
PFA coating provides additional safety- no glass or contamination if the thermometer breaks
  • The filling and glass are contained within the PFA jacket, preventing contamination
  • Enviro-Safe liquid within the thermometers is non-toxic, non-hazardous, and fully biodegradable
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy included
Double-safe thermometers are PFA safety coated In the event of breakage the coating will help contain the broken liquid and glass. Each thermometer features a black liquid and yellow back glass which creates a distinct contrast making this thermometer very easy to read. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, ring top anti-roll fitting, individual serial numbered and have a statement of accuracy.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Overall Length (mm)
Availability Pricing
H-B Instrument B60400-0300 Double-Safe PFA Coated, Liquid-In-Glass Thermometer; 20/300F, 50mm Immersion, Environmentally Friendly WW-08077-11 Mfr # 604000000
COMPARE
300 20 200
Call For Price
H-B Instrument B60400-0900 Double-Safe PFA Coated, Liquid-In-Glass Thermometer; 0/230F, 76mm Immersion, Environmentally Friendly WW-08077-17 Mfr # 604000900
COMPARE
230 0 300
Call For Price
H-B Instrument B60400-1000 Double-Safe PFA Coated, Liquid-In-Glass Thermometer; 0/300F, 76mm Immersion, Environmentally Friendly WW-08077-19 Mfr # 604001000
COMPARE
300 0 300
Call For Price
H-B Instrument B60400-2100 Double-Safe PFA Coated, Liquid-In-Glass Thermometer; 0/300F, Total Immersion, Environmentally Friendly WW-08077-26 Mfr # 604002100
COMPARE
300 0 300