H-B Instrument B60400-0900 Double-Safe PFA Coated, Liquid-In-Glass Thermometer; 0/230F, 76mm Immersion, Environmentally Friendly

Representative image only.

H-B Instrument B60400-0900 Double-Safe PFA Coated, Liquid-In-Glass Thermometer; 0/230F, 76mm Immersion, Environmentally Friendly

H-B Instrument | Mfr: B604000900| Item: 0807717

PFA coating provides additional safety- no glass or contamination if the thermometer breaks

Call For Price

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Call For Price

Buy It Together

Current Item

H-B Instrument B60400-0900 Double-Safe PFA Coated, Liquid-In-Glass Thermometer; 0/230F, 76mm Immersion, Environmentally Friendly
H-B Instrument B60400-0900 Double-Safe PFA Coated, Liquid-In-Glass Thermometer; 0/230F, 76mm Immersion, Environmentally Friendly
0807717

Call For Price
+ =
Total Price 33100.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • The filling and glass are contained within the PFA jacket, preventing contamination
  • Enviro-Safe liquid within the thermometers is non-toxic, non-hazardous, and fully biodegradable
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy included
  • 5 year warranty

More About this Item

Double-safe thermometers are PFA safety coated In the event of breakage the coating will help contain the broken liquid and glass. Each thermometer features a black liquid and yellow back glass which creates a distinct contrast making this thermometer very easy to read. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, ring top anti-roll fitting, individual serial numbered and have a statement of accuracy.

5 Year Warranty
Specifications
  • Min Temperature (° F)0
  • Length (")12
  • Max Temperature (° F)230
  • Accuracy±2.0°F
  • Division (° F)2
  • Thermometer FillBlack Spirit
  • Overall Length (InchShort)12
  • Overall Length (mm)300
  • Immersion Depth (mm)76
  • Stem MaterialPFA-Coated Yellow-Back Glass
  • DescriptionDouble-Safe PFA Coated, Liquid-In-Glass Thermometer; 0/230F, 76mm Immersion, Environmentally Friendly
  • Warranty5 years
Reviews
Q&A