Guardian Equipment Eyewash/Drench Hose Units
Meet ANSI standards for both eyewashes and drench hoses
  • Provides a soft, balanced flow of water
  • Built-in filters remove debris
  • ANSI Z358.1-2004 compliance
Combination eyewash/drench hose units have two GS-plus sprayheads. All sprayheads feature self-regulating flow control valves to provide a soft, balanced flow of water and built-in filters remove debris from water. Eyewash/drench hose units are 100% factory assembled and tested.
Compliance: ANSI Z358.1-2004
103,000.00 - ₹131,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Eyewash Flow Rate (GPM) Hose Length (ft) Hose Material
Availability Pricing
Guardian Equipment G5026 Wall-Mount Eyewash/Drench Hose Unit with Squeeze-Open Valve 0676712 Mfr # G5026
COMPARE
1.8 8 Reinforced PVC
₹103,000.00INR / Each
Guardian Equipment G5014 Wall-Mount Eyewash/Drench Hose Unit with Stay-Open Valve 0676714 Mfr # G5014
COMPARE
3.6 12 Nylon
₹131,000.00INR / Each
Guardian Equipment G5022 Counter top mounted combination eyewash/drench hose with stay-open valve 0676762 Mfr # G5022
COMPARE
3.6 8 Reinforced PVC
₹110,000.00INR / Each