Guardian Equipment Eyewash Bowls
Spray head assembly features internal flow control and filter to remove impurities from the water flow.
Sprayhead assembly has two gentle spray outlet heads with flip-top dust covers mounted on chrome-plated brass supply arms. Activate water flow with cast-aluminum flag handle attached to a stay-open valve. The 11-1⁄2"-dia bowl acts like a sink to drain water away. Units are factory tested. Supply connection is 1⁄2" NPT(F); waste connection is 1-1⁄4" NPT(M).
73,500.00 - ₹159,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bowl Material Eyewash Fixture Material Mount
Availability Pricing
Guardian Equipment G1814P Wall Mount Eye Wash, ABS plastic 0676715 Mfr # G1814P
COMPARE
ABS plastic Chrome-plated brass Wall Mount
₹73,500.00INR / Each
Guardian Equipment G1814 Wall Mount Eye Wash, Stainless Steel 0676716 Mfr # G1814
COMPARE
Stainless steel Chrome-plated brass Wall Mount
₹94,100.00INR / Each
Guardian Equipment G1814BC Wall Mount Eye Wash, Stainless Steel bowl with cover 0676717 Mfr # G1814BC
COMPARE
Stainless steel Chrome-plated brass Wall Mount
₹159,000.00INR / Each
Guardian Equipment G1825HFC Pedestal Eye Wash, Stainless Steel bowl with foot petal 0676719 Mfr # G1825HFC
COMPARE
Stainless steel Chrome-plated brass Pedestal Mount
₹146,000.00INR / Each