Guardian Equipment G1814 Wall Mount Eye Wash, Stainless Steel
Representative image only.
Guardian Equipment G1814 Wall Mount Eye Wash, Stainless Steel
Guardian Equipment | Mfr: G1814| Item: 0676716
Spray head assembly features internal flow control and filter to remove impurities from the water flow.
₹94,100.00INR / Each
₹94,100.00INR / Each
Product Description
Includes
ANSI-compliant identifcation sign
Key Features
  • 2 year warranty
More About this Item
Sprayhead assembly has two gentle spray outlet heads with flip-top dust covers mounted on chrome-plated brass supply arms. Activate water flow with cast-aluminum flag handle attached to a stay-open valve. The 11-1⁄2"-dia bowl acts like a sink to drain water away. Units are factory tested. Supply connection is 1⁄2" NPT(F); waste connection is 1-1⁄4" NPT(M).
Wall-mount units include mounting brackets.
Compliance: ANSI Z385.1-2004
Two year warranty
Specifications
  • Eyewash Flow Rate (GPM)3.6
  • Eyewash Fixture MaterialChrome-plated brass
  • Bowl MaterialStainless steel
  • Supply Pipe MaterialChrome-plated brass
  • Waste Pipe MaterialCast aluminum
  • MountWall Mount
  • Outlet Connections1-1/4" NPT(F)
  • Inlet Connection1/2" NPT(F)
  • ComplianceANSI Z385.1-2009
  • DescriptionWall Mount Eye Wash, Stainless Steel