Guardian Equipment Deck-Mount AutoFlow™ Personal Eyewash
Swing spray heads over sink to activate waterflow with AutoFlow™
  • Constructed of durable chrome-plated brass
  • Two GS-Plus™ spray heads with “flip-top” dust cover provide flow control and filter to remove impurities from water
  • Easily visible ANSI-compliant identification sign
These Guardian Equipment deck-mount AutoFlow™ eyewash stations allow for easy mounting on counter next to a sink. AutoFlow™ allows spray to automatically activate when eyewash arm is swung over a sink or pulled down; water flow continues until the spray head assembly is moved back to its original position. Eyewash stations are furnished with an in-line strainer to protect the valve and spray heads from debris in the water line. All units are completely assembled and water tested prior to shipment.
Choose from the 90° right hand swivel mount or 90° swing down mount versions.
114,000.00 - ₹183,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Eyewash Fixture Material Mount Description
Availability Pricing
Guardian Equipment Deck-Mount AutoFlow™ Personal Eyewash, 90° Swivel, Right Hand Mount 0185055 Mfr # G1805
COMPARE
Chrome-Plated Brass Deck/Counter, Right Side Deck-Mount AutoFlow™ Personal Eyewash, 90° Swivel, Right Hand Mount
₹114,000.00INR / Each
Guardian Equipment Deck-Mount AutoFlow™ Personal Eyewash, 90° Swing Down 0185056 Mfr # G1849
COMPARE
Chrome-Plated Brass Deck/Counter Deck-Mount AutoFlow™ Personal Eyewash, 90° Swing Down
₹183,000.00INR / Each