Guardian Equipment Deck-Mount AutoFlow™ Personal Eyewash, 90° Swivel, Right Hand Mount
Representative image only.
Guardian Equipment Deck-Mount AutoFlow™ Personal Eyewash, 90° Swivel, Right Hand Mount
Guardian Equipment | Mfr: G1805| Item: 0185055
Swing spray heads over sink to activate waterflow with AutoFlow™
₹114,000.00INR / Each
₹114,000.00INR / Each
Product Description
Key Features
  • Constructed of durable chrome-plated brass
  • Two GS-Plus™ spray heads with “flip-top” dust cover provide flow control and filter to remove impurities from water
  • Easily visible ANSI-compliant identification sign
  • Swinging the spray head assembly 90° horizontally out over the sink activates the water flow
More About this Item
These Guardian Equipment deck-mount AutoFlow™ eyewash stations allow for easy mounting on counter next to a sink. AutoFlow™ allows spray to automatically activate when eyewash arm is swung over a sink or pulled down; water flow continues until the spray head assembly is moved back to its original position. Eyewash stations are furnished with an in-line strainer to protect the valve and spray heads from debris in the water line. All units are completely assembled and water tested prior to shipment.
Choose from the 90° right hand swivel mount or 90° swing down mount versions.
Specifications
  • Eyewash Fixture MaterialChrome-Plated Brass
  • Supply Pipe MaterialChrome-Plated Brass
  • MountDeck/Counter, Right Side
  • Inlet Connection1/2" NPT(F)
  • ComplianceANSI Z385.1-2009
  • DescriptionDeck-Mount AutoFlow™ Personal Eyewash, 90° Swivel, Right Hand Mount