Grieve NB Series Industrial Mechanical Convection Bench Ovens
Durable stainless-steel construction for harsh environments
  • Interior surface is highly resistant to corrosion and oxidation
  • Double doors allow easy loading of baskets and large parts
  • Calibrated temperature controller
  • Holds a maximum of 2 shelves
These economical mechanical convection bench ovens allow you to easily move them anywhere in your lab where needed. On-off panel switch controls power to the unit and a red pilot light indicates when the heater is on. Incoloy® sheathed heating elements and the stainless-steel interior are highly resistant to corrosion and oxidation. High-density rock wool insulation effectively retains heat for improved temperature uniformity. Double doors offer greater access to the chamber—ideal when loading larger items.
Overtemperature protection turns oven off if unit exceeds maximum temperature; simply press button to reset oven. Other features include recirculating blower, silicone rubber door gasket, and brushed stainless steel exterior.
936,000.00 - ₹1,770,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Max Temperature (° F) Power (VAC)
Availability Pricing
Grieve NB-350 Industrial Mechanical Convection Bench Oven, 7 cu ft; 230V 5230205 Mfr # NB350-230V/60HZ
COMPARE
7 350 230
₹936,000.00INR / Each
Grieve NB-550 Industrial Mechanical Convection Bench Oven, 5.3 cu ft; 115V 5230206 Mfr # NB550-115V/60HZ
COMPARE
5.3 550 115
₹1,665,000.00INR / Each
Grieve NB-550 Industrial Mechanical Convection Bench Oven, 5.3 cu ft; 230V 5230207 Mfr # NB550-230V/60HZ
COMPARE
5.3 550 230
₹1,770,000.00INR / Each