Glass Tempering Beakers
Maintain constant sample temperatures
  • Connect circulator, fill with sample and maintain a constant temperature—it’s that easy
  • Tubing connectors are 1-1⁄2"L x 1⁄2" OD
Liquid-jacketed borosilicate glass vessels have double-walled casing for circulating cold or warm tempering fluid. Tubing connectors measure 1 1⁄2"L x 1⁄2" OD. Connect a constant temperature circulator to maintain sample temperatures.
55,500.00 - ₹147,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
COMPARE
250 25 Glass
₹55,500.00INR / Each
COMPARE
600 50 Glass
₹71,400.00INR / Each
COMPARE
2000 100 Glass
₹147,000.00INR / Each