Glass Cold Traps
Cold traps are used with Dewar flasks and ice/liquid N2 to prevent condensable solvents from damaging vacuum pumps. Borosilicate glass traps withstand full vacuum.
75,200.00 - ₹112,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Pyrex 7729-28 Cold trap, glass, 40 mL, 10 mm OD WW-34530-30 Mfr # 7729-28
COMPARE
Cold trap, glass, 40 mL, 10 mm OD
₹112,000.00INR / Each
Pyrex 7729-38 Cold trap, glass, 70 mL, 16 mm OD WW-34530-34 Mfr # 7729-38
COMPARE
Cold trap, glass, 70 mL, 16 mm OD
₹75,200.00INR / Each