Pyrex 7729-28 Cold trap, glass, 40 mL, 10 mm OD

Representative image only.

Pyrex 7729-28 Cold trap, glass, 40 mL, 10 mm OD

Pyrex | Mfr: 7729-28| Item: 3453030

₹51,700.00INR / Each

₹51,700.00INR / Each
Product Description

More About this Item

Cold traps are used with Dewar flasks and ice/liquid N2 to prevent condensable solvents from damaging vacuum pumps. Borosilicate glass traps withstand full vacuum.

Specifications
  • DescriptionCold trap, glass, 40 mL, 10 mm OD
Reviews
Q&A