Glas-Col Stirring/Heating Mantle Silicone Spillproof Guards
Prevents damage from chemical spillage
  • Improves operating safety when heating liquids
  • Rugged silicone pad holds guard securely in place
These spillproof guards prevent mantle damage from chemical spillage and improve operating safety when heating liquids. The guard is appropriate for use with low-wattage mantles for flask. A rugged silicone pad grips outer circumference of vessel, holding the guard securely in place.
60,800.00 - ₹99,800.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flask Size (mL) Description
Availability Pricing
Glas-Col 100D SI05 Stirring/Heating Mantle Silicone Spillproof Guard; 5L 3622694 Mfr # 100D SI05
COMPARE
5000 Stirring/Heating Mantle Silicone Spillproof Guard; 5L
₹60,800.00INR / Each
Glas-Col 100D SI12 Stirring/Heating Mantle Silicone Spillproof Guard; 12L 3622695 Mfr # 100D SI12
COMPARE
12000 Stirring/Heating Mantle Silicone Spillproof Guard; 12L
₹73,500.00INR / Each
Glas-Col 100D SI22 Stirring/Heating Mantle Silicone Spillproof Guard; 22L 3622696 Mfr # 100D SI22
COMPARE
22000 Stirring/Heating Mantle Silicone Spillproof Guard; 22L
₹99,800.00INR / Each