Glas-Col Stirring/Heating Mantle Fabric Insulating Tops
Prevents vapor condensation
  • Provides added insulation and protect
The fabric insulating tops prevent vapor condensation in the upper half of a flask during distillation of high-boiling materials. It also protects the flask, solutions, and laboratory from drafts, flying glass of flask implosions in high vacuum distillations, falling objects, and thermal strain between top and lower half of flask during high-temperature operations. The glass-fabric construction, laced-side opening, and elliptical hole accomodates 1-, 2-, and 3-neck flasks.
86,100.00 - ₹202,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flask Size (mL) Description
Availability Pricing
Glas-Col 100A INS1103 Stirring/Heating Mantle Fabric Insulating Top; 2L 3622680 Mfr # 100A INS1103
COMPARE
2000 Stirring/Heating Mantle Fabric Insulating Top; 2L
₹87,800.00INR / Each
Glas-Col 100A INS1123 Stirring/Heating Mantle Fabric Insulating Top; 3L 3622681 Mfr # 100A INS1123
COMPARE
3000 Stirring/Heating Mantle Fabric Insulating Top; 3L
₹86,100.00INR / Each
Glas-Col 100A INS1143 Stirring/Heating Mantle Fabric Insulating Top; 5L 3622682 Mfr # 100A INS1143
COMPARE
5000 Stirring/Heating Mantle Fabric Insulating Top; 5L
₹117,000.00INR / Each
Glas-Col 100A INS1163 Stirring/Heating Mantle Fabric Insulating Top; 12L 3622683 Mfr # 100A INS1163
COMPARE
12000 Stirring/Heating Mantle Fabric Insulating Top; 12L
₹177,000.00INR / Each
Glas-Col 100A INS1183 Stirring/Heating Mantle Fabric Insulating Top; 22L 3622684 Mfr # 100A INS1183
COMPARE
22000 Stirring/Heating Mantle Fabric Insulating Top; 22L
₹202,000.00INR / Each