Glas-Col Aluminum Flask Heating Mantles
Get uniform temperature control for low-volume samples
  • Rigid housing provides strength and durability while supporting the vessel weight
  • Fabric interior minimizes thermal shock and damage to glassware
  • Maximum temperature of 842°F (450°C)
Flask mantles feature durable aluminum exterior and soft fabric interior. Mantles cover the bottom half of the flask allowing full view of the contents inside. Larger sizes have multiple circuits for ease of temperature control when a flask is less than half full. Maximum temperature is 842°>F (450°>C). Mantles require a controlled electrical power source to prevent overheating.
140,000.00 - ₹318,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flask Size (mL) Power (VAC)
Availability Pricing
Glas-Col 100B TM109 Aluminum Heating Mantle, 1000 ml, 380 Watts, 230 V 3623326 Mfr # 100B TM109CE
COMPARE
1000 230
₹140,000.00INR / Each
Glas-Col 100B TM111 Aluminum Heating Mantle, 2000 ml, 500 Watts, 230 V 3623336 Mfr # 100B TM111CE
COMPARE
2000 230
₹143,000.00INR / Each
Glas-Col 100B TM113 Aluminum Heating Mantle, 3000 ml, 500 Watts, 230 V 3623346 Mfr # 100B TM113CE
COMPARE
3000 230
₹186,000.00INR / Each
Glas-Col 100B TM115 Aluminum Heating Mantle, 5000 ml, 600 Watts, 230 V 3623356 Mfr # 100B TM115CE
COMPARE
5000 230
₹214,000.00INR / Each
Glas-Col 100B TM117 Aluminum Heating Mantle, 12,000 ml, 2@650 Watts, 230 V 3623366 Mfr # 100B TM117CE
COMPARE
12000 230
₹318,000.00INR / Each