Foxx Life Sciences Storage Carboys w/Spigot, Polypropylene
Secure grips make lifting, filling, and pouring easy
  • Easy-to-read metric graduations are certified to ±5% accuracy
  • Wide-mouth neck simplifies filling and cleaning
  • Ergonomic body design maximizes bench and shelf space
  • Large top handles offer full hand grip
  • Interchangeable leak-proof top connections
  • BPA-free, USP Class VI, FDA-grade autoclavable resin offers chemical resistance and heat stability
The easy-to-grip polypropylene storage carboys allow you to securely hold the carboy while lifting, filling, and pouring. The carboys feature easy-to-read metric graduation marks and molded-in material and volume identification markings. Include autoclavable PP spigot.
28,800.00 - ₹49,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Special Features
Availability Pricing
Foxx Life Sciences Carboy, Polypropylene, 83 mm Cap, Spigot; 5 L 0605951 Mfr # 155-1211-OEM
COMPARE
5 80 mm Spigot
₹28,800.00INR / Each
Foxx Life Sciences Carboy, Polypropylene, 83 mm Cap, Spigot; 10 L 0605954 Mfr # 155-2211-OEM
COMPARE
10 80 mm Spigot
₹36,000.00INR / Each
Foxx Life Sciences Carboy, Polypropylene, 83 mm Cap, Spigot; 20 L 0605958 Mfr # 155-3211-OEM
COMPARE
20 80 mm Spigot
₹46,400.00INR / Each
Foxx Life Sciences Carboy, Polypropylene, 120 mm Cap, Spigot; 20 L 0605959 Mfr # 155-3221-OEM
COMPARE
20 120 mm Spigot
₹49,000.00INR / Each