FLIR MR-Series Thermal Imaging Moisture Meters
Take more confident moisture readings
  • Built-in 80 x 60 thermal imager with IGM scanning technology
  • Document thermal images and moisture readings on screen
  • Review images and generate reports with FLIR Tools software
  • Pinless technology for fast noncontact measurements
  • External pin probe for more precise direct-contact moisture measurements
  • Internal rechargeable battery
These versatile imaging moisture meters have a built-in thermal camera—Infrared Guided Measurement (IGM) technology—to help you quickly scan and target moisture issues. The live thermal images visually guide you to problem areas to make more confident moisture readings, and the integrated laser and crosshair let you easily reference problem areas.
Once the optimal testing location is located, the meters measure the moisture content with either an integrated pinless sensor or an external pin probe—providing flexibility to take either nonintrusive or intrusive measurements before, during, and after the drying process. Optional pin probes allow you to test in challenging areas, expanding the functionality of your meter.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Measurement Type Description
Availability Pricing
Flir MR176 IGM™  Moisture Meter with Replaceable Hygrometer 3780213 Mfr # MR176
COMPARE
1 3.7V rechargeable lithium-ion (included) Moisture IGM™ Moisture Meter with Replaceable Hygrometer
Call For Price
Flir MR-Series  Imaging Moisture Meter Kit Combines MR160 + MR08 Hammer & Wall Probe 3975615 Mfr # MR160-KIT5
COMPARE
3.7V Rechargeable Moisture Imaging Moisture Meter Kit Combines MR160 + MR08 Hammer & Wall Probe
Call For Price
Flir MR-Series  Imaging Moisture Meter Kit Combines MR176 + MR08 Hammer & Wall Probe 3975616 Mfr # MR176-KIT5
COMPARE
1 3.7V rechargeable lithium-ion (included) Moisture Imaging Moisture Meter Kit Combines MR176 + MR08 Hammer & Wall Probe
Call For Price
Flir MR-Series  Imaging Moisture Meter Kit Combines MR160 + MR08 Hammer Probe & C2 Thermal Imager 3975675 Mfr # MR160-KIT2
COMPARE
3.7V Rechargeable Moisture Imaging Moisture Meter Kit Combines MR160 + MR08 Hammer Probe & C2 Thermal Imager
Call For Price
Flir MR-Series  Imaging Moisture Meter Kit Combines MR176 + MR08 Hammer Probe & E6 Thermal Imager 3975676 Mfr # MR176-KIT6
COMPARE
1 3.7V rechargeable lithium-ion (included) Moisture Imaging Moisture Meter Kit Combines MR176 + MR08 Hammer Probe & E6 Thermal Imager