Fixed-Volume Lightweight Pipettors, Metal
Compact size and durability makes pipettes ideal for field work
  • Highly accurate pipettors with durable, yet lightweight, metal construction
  • One-stroke plunger movement provides precise results
  • Fully Autoclavable to avoid cross-contamination
  • Color coded by volumes for easy identification
For accurate and consistent results, choose a fixed-volume pipettor. Ideal for repetitive delivery of reagents and solutions, these durable lightweight pipettors feature a precision-machined plunger and barrel assembly. The internal polyurethane multi-ring seals prevent fluid leakage far superior to single O-ring pipettors. Pipettors are color-coded to help you keep track of the proper dispensing volume — an excellent choice for a busy lab!
33,900.00 - ₹58,600.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy (%) Color Code Max Volume (µL)
Availability Pricing
COMPARE
±0.70% Brown 25
₹38,600.00INR / Each
COMPARE
±0.40% Red 100
₹36,600.00INR / Each
COMPARE
±0.20% Red 1000
₹33,900.00INR / Each
COMPARE
±0.10% Yellow 2000
₹58,600.00INR / Each
COMPARE
±0.10% Blue 5000
₹41,600.00INR / Each