Fixed-Volume Lightweight Pipettor, Metal, Blue, 5000 µL
Representative image only.
Fixed-Volume Lightweight Pipettor, Metal, Blue, 5000 µL
| Item: 0783976
Compact size and durability makes pipettes ideal for field work
₹41,600.00INR / Each
for an additional
₹14,500.00INR / Each
for an additional
₹25,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹41,600.00INR / Each
Buy It Together
Current Item
Fixed-Volume Lightweight Pipettor, Metal, Blue, 5000 µL
Fixed-Volume Lightweight Pipettor, Metal, Blue, 5000 µL
0783976
₹41,600.00INR / Each
+ =
Total Price 56100.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Highly accurate pipettors with durable, yet lightweight, metal construction
  • One-stroke plunger movement provides precise results
  • Fully Autoclavable to avoid cross-contamination
  • Color coded by volumes for easy identification
  • 1 year warranty
More About this Item
For accurate and consistent results, choose a fixed-volume pipettor. Ideal for repetitive delivery of reagents and solutions, these durable lightweight pipettors feature a precision-machined plunger and barrel assembly. The internal polyurethane multi-ring seals prevent fluid leakage far superior to single O-ring pipettors. Pipettors are color-coded to help you keep track of the proper dispensing volume — an excellent choice for a busy lab!
1 Year Warranty
Specifications
  • SterileNo
  • AutoclavableYes
  • Max Volume (µL)5000
  • Accuracy (%)±0.10%
  • Reproducibility±0.12%
  • Overall Length (InchShort)8
  • Overall Length (mm)203.2
  • Number Of Channels1
  • Tip EjectorNo
  • Body MaterialMetal
  • Color CodeBlue
  • DescriptionFixed-Volume Lightweight Pipettor, Metal, Blue, 5000 µL