Extech PRC10 and PRC15 Precision Current and Precision Current/Voltage Calibrators
Large battery bank for extended current and voltage calibration
  • Palm-sized double molded housing
  • Large dot-matrix digital backlit LCD
  • Standard banana I/O ports
  • Up to five user adjustable calibration presets
  • Auto power off feature
  • External power adapter for continuous work cycle
130,000.00 - ₹133,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Measurement (mA) Max Temperature (° F) Description
Availability Pricing
Extech PRC10 Current Calibrator and Meter 1610113 Mfr # PRC10
COMPARE
50 41 Current Calibrator and Meter
₹133,000.00INR / Each
Extech PRC15 Current and Voltage Calibrator and Meter 1610114 Mfr # PRC15
COMPARE
50 41 Current and Voltage Calibrator and Meter
₹130,000.00INR / Each