Extech Data Loggers with SD Memory Card
SD Card allows you to remove past data from unit and analyze while monitoring simultaneously
Use these digital data loggers to monitor food, chemical, drug, and equipment storage areas, cleanrooms, incubators, and more! The easy-to-read LCD allows you to view current readings while recording. Set the flexible data logging interval to collect sample data from 5 seconds to 10 minutes. Data logging software stamps and stores date/time readings on an SD card in an Excel® format for easy transfer to a PC. These data loggers can be programmed to set-off alarms at preset values providing warning when critical enviromental factors pose jeopardy to processes, goods, or people. The compact rugged design allows you to place these data loggers just about anywhere.
56,700.00 - ₹96,100.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Temperature Accuracy (° F) Temperature Resolution (° F)
Availability Pricing
Extech SD200 3-Channel Temperature Data Logger WW-70001-30 Mfr # SD200
COMPARE
2372 ±0.9 0.1
₹58,800.00INR / Each
Extech SD500 Temperature/Humidity Data Logger WW-70001-31 Mfr # SD500
COMPARE
122 ±1.8 0.1
₹56,700.00INR / Each
Extech SD800 Temperature/Humidity/CO2 Data Logger WW-70001-32 Mfr # SD800
COMPARE
122 ±0.45 0.1
₹96,100.00INR / Each