Excel Scientific X-Pierce Sealing Film
Precut ''X'' over each well ideal for automated sampling
  • Film reseals after sampling
  • Functional temperature -40 to 90°C
These 100-µm thick vinyl films are designed for temporary protection from contamination and evaporation for samples in 96-well plates. A precut ''X'' over each well creates four flaps that easily bend downward when pushed by a robotic probe or pipet tip, allowing sample access without coring or adhesive fouling. For long-term protection after sampling, a continuous film should be applied as a second layer. Measures 79.4 x 145.5-mm.
18,500.00 - ₹29,500.00INR / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Sterile Description
Availability Pricing
Excel Scientific Precut Piercable Sealing Films, 100/pk 1302440 Mfr # XP-100
COMPARE
No Precut Piercable Sealing Films, 100/pk
₹29,500.00INR / Pkg of 100
Excel Scientific Precut Piercable Sealing Films, 50/pk - Sterile 1302442 Mfr # XPS-25
COMPARE
Yes Precut Piercable Sealing Films, 50/pk - Sterile
₹18,500.00INR / Pkg of 50