Excel Scientific Precut Piercable Sealing Films, 50/pk - Sterile
Representative image only.
Excel Scientific Precut Piercable Sealing Films, 50/pk - Sterile
Excel Scientific | Mfr: XPS-25| Item: 1302442
Precut ''X'' over each well ideal for automated sampling
₹18,500.00INR / Pkg of 50
₹18,500.00INR / Pkg of 50
Product Description
Key Features
  • Film reseals after sampling
  • Functional temperature -40 to 90°C
  • 3 year warranty
More About this Item
These 100-µm thick vinyl films are designed for temporary protection from contamination and evaporation for samples in 96-well plates. A precut ''X'' over each well creates four flaps that easily bend downward when pushed by a robotic probe or pipet tip, allowing sample access without coring or adhesive fouling. For long-term protection after sampling, a continuous film should be applied as a second layer. Measures 79.4 x 145.5-mm.
3 Year Warranty
Specifications
  • SterileYes
  • Plate TypeSealing film
  • DescriptionPrecut Piercable Sealing Films, 50/pk - Sterile