Etatron DLXB Series Foot-Mount pH/ORP Pump Control Systems
Integrated controller and metering pump simplify system setup
  • Control systems are able to accept pH or ORP electrodes for added convenience
  • On/off or proportional control modes allow for robust process control
  • Deadband is programmable for greater flexibility
  • LCD is easy to read
Control the pH or ORP levels of your process with an integrated controller and pump—saves you time, money, and space. These solenoid-driven, diaphragm metering pumps and controllers are encased with an IP65, acid-resistant plastic housing. Polypropylene pump heads have manual air bleed, PTFE diaphragm, and Viton wetted parts. Pump control systems accept level switches (order separately). Backlit LCD display has resolution of 0.01 pH or 1 mV.
Controllers are programmable for on/off or proportional control applications and have manual temperature compensation. Control can be fine-tuned by programmable hysteresis and delay functions. Frequency of injections is programmable between 0 to 100% of the maximum flow rate of the pump.
Control systems accept electrodes with BNC connectors and no temperature compensation; order electrodes separately.
See instruction manual for complete specifications
188,000.00 - ₹251,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPH) Max Pressure (PSI) Power (VAC)
Availability Pricing
Etatron DLXB-PH-RX/MBB DLXB pH/ORP Pump Control System, 1 L/hr, 15 bar, Foot-Mt; 115V 5602247 Mfr # PBX3622110EA
COMPARE
0.26 217.5 115
₹189,000.00INR / Each
Etatron DLXB-PH-RX/MBB DLXB pH/ORP Pump Control System, 5 L/hr, 7 bar, Foot-Mt; 115V 5602249 Mfr # PBX3608610EA
COMPARE
1.32 101.5 115
₹188,000.00INR / Each
Etatron DLXB-PH-RX/MBB DLXB pH/ORP Pump Control System, 8 L/hr, 10 bar, Foot-Mt; 115V 5602251 Mfr # PBX3622710EA
COMPARE
2.11 145 115
₹201,000.00INR / Each
Etatron DLXB-PH-RX/MBB DLXB pH/ORP Pump Control System, 15 L/hr, 4 bar, Foot-Mt; 115V 5602252 Mfr # PBX3623010EA
COMPARE
3.96 58 115
₹239,000.00INR / Each
Etatron DLXB-PH-RX/MBB DLXB pH/ORP Pump Control System, 20 L/hr, 3 bar, Foot-Mt; 115V 5602253 Mfr # PBX3609510EA
COMPARE
5.28 43.5 115
₹227,000.00INR / Each
Etatron DLXB-PH-RX/MBB DLXB pH/ORP Pump Control System, 1 L/hr, 15 bar, Foot-Mt; 230V 5602287 Mfr # PBX3622210EA
COMPARE
0.26 217.5 230
₹188,000.00INR / Each
Etatron DLXB-PH-RX/MBB DLXB pH/ORP Pump Control System, 15 L/hr, 4 bar, Foot-Mt; 230V 5602292 Mfr # PBX3623110EA
COMPARE
3.96 58 230
₹239,000.00INR / Each
Etatron DLXB-PH-RX/MBB DLXB pH/ORP Pump Control System, 20 L/hr, 3 bar, Foot-Mt; 230V 5602293 Mfr # PBX3622010EA
COMPARE
5.28 43.5 230
₹251,000.00INR / Each
Showing 1- 8 of 8