Eppendorf® Xplorer® Electronic Pipettors
Superior design provides optimal support of your work with industry-leading accuracy and features
  • Precise adjustable parameters of all functions with the practical selection dial
  • Intuitive user interface and organized color display
  • Universal nose cone accepts most pipette tips
  • Lightweight design and optimal readability in every position offers unsurpassed ergonomics
  • Multilingual operation
The Eppendorf Xplorer electronic pipettor was specially designed for high professional standards with a new intuitive operating concept and design of simplicity, precision and reproducibility. The selection dial provides all functions in plain view and eliminates delays due to complicated programming. The modern color display is easy to use and does not have any submenus. The multifunction rocker offers industry-leading technology, so your pipettor is always readable, no matter how you handle or hold it.
The innovative ejector automatically returns to the zero position after tip ejection. A powerful battery offers long-lasting use and you may charge the pipettor separately. Unit will automatically shut off while not in use.
The multi-function rocker offers industry leading technology, so your pipettor is always readable, no matter how you handle it. Choose from various languages to use your pipettor.
Call For Price
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Min Volume (µL) Number Of Channels
Availability Pricing
Eppendorf 4861000023 Single-Channel Digital Pipette w/ Charger; 5-100 µL WW-24500-69 Mfr # 4861000023
COMPARE
100 5 1
Call For Price
Eppendorf 4861000031 Single-Channel Digital Pipette w/ Charger; 15-300 µL WW-24500-73 Mfr # 4861000031
COMPARE
300 15 1
Call For Price
Eppendorf 4861000040 Single-Channel Digital Pipette w/ Charger; 50-1000 µL WW-24500-74 Mfr # 4861000040
COMPARE
1000 50 1
Call For Price
Eppendorf 4861000058 Single-Channel Electronic Pipette (incl.charging), 0.25-5mL WW-24500-78 Mfr # 4861000058
COMPARE
5000 250 1
Call For Price
Eppendorf 4861000066 Single-Channel Electronic Pipette (incl.charging), 0.5-10mL WW-24500-79 Mfr # 4861000066
COMPARE
10000 500 1
Call For Price
Eppendorf 4861000104 8-Channel Digital Pipette w/ Charger; 0.5-10 µL WW-24500-80 Mfr # 4861000104
COMPARE
10 0.5 8
Call For Price
Eppendorf 4861000120 8-Channel Digital Pipette w/ Charger; 5-100 µL WW-24500-81 Mfr # 4861000120
COMPARE
100 5 8
Call For Price
Eppendorf 4861000147 8-Channel Digital Pipette w/ Charger; 15-300 µL WW-24500-82 Mfr # 4861000147
COMPARE
300 15 8
Call For Price
Eppendorf 4861000163 8-Channel Digital Pipette w/ Charger; 50-1200 µL WW-24500-83 Mfr # 4861000163
COMPARE
1200 50 8
Call For Price
Eppendorf 4861000155 12-Channel Digital Pipette w/ Charger; 15-300 µL WW-24500-89 Mfr # 4861000155
COMPARE
300 15 12
Call For Price
Showing 1- 10 of 10