Eppendorf epTIPS PCR Clean Pipette Tips
Refill system and reduced ejection force requirement
The Eppendorf epTIPS - Totally Integrated Pipetting System – satisfiy the stringent requirements of dependability, reproducibility, and the trend towards smaller sample volumes.
Optimized cone geometry allows for a perfect matching of every Eppendorf pipette tip to every Eppendorf pipette, for guaranteed pipette performance when used as a system. Additionally, the epTIPS are also universally applicable for use with pipettes from all other manufacturers.
The epTIPS system provides an alternative to throwing away an entire box when the tips run out. The Reload set enables contamination-free transfer of fresh tip trays into the reusable box used for loading your pipettor. For one-time use tip boxes, order a Rack.
PCR clean tips are certified human DNA-free, DNase-free, RNase-free, and PCR inhibitor-free.
23,100.00 - ₹29,100.00INR / Case of 960
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Min Volume (µL) Package Of
Availability Pricing
Eppendorf 022491709 PCR Clean Pipette Tips, 0.1 to 10 uL, 10 reloads, 960/cs WW-18888-82 Mfr # 022491709
COMPARE
10 0.1 10 Reloads; 960/cs
₹29,100.00INR / Case of 960
Eppendorf 022491717 PCR Clean Pipette Tips, 0.1 to 20 uL, 10 reloads, 960/cs WW-18888-84 Mfr # 022491717
COMPARE
20 0.1 10 Reloads; 960/cs
₹29,100.00INR / Case of 960
Eppendorf 022491725 PCR Clean Pipette Tips, 0.5 to 20 uL, 10 reloads, 960/cs WW-18888-86 Mfr # 022491725
COMPARE
20 0.5 10 Reloads; 960/cs
₹29,100.00INR / Case of 960
Eppendorf 022491733 PCR Clean Pipette Tips, 2 to 200 uL, 10 reloads, 960/cs WW-18888-88 Mfr # 022491733
COMPARE
200 2 10 Reloads; 960/cs
₹23,100.00INR / Case of 960
Eppendorf 022491768 PCR Clean Pipette Tips, 50 to 1250 uL, 10 reloads, 960/cs WW-18888-94 Mfr # 022491768
COMPARE
1250 50 10 Reloads; 960/cs
₹25,000.00INR / Case of 960