Environmental Express® Straight-Sided Wide-Mouth Jars, HDPE or PP
Gain easier access to your samples
  • Large diameter lid allows for easier scooping and removal of samples
  • Reach entire sample with straight-sided design
  • Level 1 containers are lot controlled, sealed with custody tape, and include a Certificate of Analysis from an independent laboratory
  • Level 3 containers meet EPA guidelines, but do not ship with a certificate of analysis
Included leakproof caps are composed of polypropylene (PP) and lined with F217, a low-density foam core between two solid layers of LDPE.
6,300.00 - ₹29,700.00INR / Case of 15
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Description
Availability Pricing
COMPARE
250 Polypropylene(PP) Straight-Sided Wide-Mouth Jar, PP, Lev 1, 250 mL; 24/Cs
₹6,300.00INR / Case of 24
COMPARE
250 Polypropylene(PP) Straight-Sided Wide-Mouth Jar, Polypropylene (PP), Level 3, 250 mL; 120/CS
₹29,700.00INR / Case of 120
COMPARE
500 High Density Polyethyene (HDPE) Straight-Sided Wide-Mouth Jar, HDPE, Level 1, 500 mL; 15/CS
₹8,100.00INR / Case of 15
COMPARE
500 Polypropylene(PP) Straight-Sided Wide-Mouth Jar, PP, Lev 1, 500 mL; 24/Cs
₹9,500.00INR / Case of 24
COMPARE
1000 High Density Polyethyene (HDPE) Straight-Sided Wide-Mouth Jar, HDPE, Level 1, 1000 mL; 12/CS
₹6,700.00INR / Case of 12
COMPARE
125 Polypropylene(PP) Straight-Sided Wide-Mouth Jar, PP, White, Level 1, 125 mL; 175/Cs
₹29,000.00INR / Case of 175
COMPARE
250 High Density Polyethyene (HDPE) Straight-Sided Wide-Mouth Jar, HDPE, Natural, Level 1, 250 mL; 120/CS
₹29,000.00INR / Case of 120
COMPARE
500 High Density Polyethyene (HDPE) Straight-Sided Wide-Mouth Jar, HDPE, Natural, Level 3, 500 mL; 24/CS
₹6,800.00INR / Case of 24
COMPARE
1000 High Density Polyethyene (HDPE) Straight-Sided Wide-Mouth Jar, HDPE, Natural, Level 3, 1000 mL; 12/CS
₹7,600.00INR / Case of 12
COMPARE
1000 High Density Polyethyene (HDPE) Straight-Sided Wide-Mouth Jar, HDPE, Natural, Level 3, 1000 mL; 84/CS
₹27,400.00INR / Case of 84
Showing 1- 10 of 10