Environmental Express StepSaver™ Individual Extraction Stations
Small footprint saves valuable benchtop space
  • Can be daisy-chained together to run multiple samples at one
  • Meets EPA method 1664 requirements for oil and grease extractions
  • Two separate chambers help keep water out of your extract
Constructed out of durable PVC and plastic materials, these compact, single-place extraction stations make oil and grease extractions easy. They are designed to be used for any type of Solid Phase Extraction (SPE) technique using an extraction disk. Single stations can be daisy-chained together to run multiple samples at once for faster sample throughput.
StepSaver system allows you to easily and efficiently collect the solvent with eluted oil and grease directly into a collection flask. No need to disassemble or reassemble glassware. As fluid enters the collection flask, it passes through a drying cartridge to remove excess water. At this point it is ready to be evaporated without a separate drying step.
The extraction stations are available in three different configurations: disposable filter funnel with UtraPrep® extraction disk, 47 mm glass filter funnel, and 90 mm glass filter funnel.
251,000.00 - ₹344,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Diameter (mm) Funnel Size (mL) Description
Availability Pricing
Environmental Express StepSaver™ Oil and Grease Extraction Individual Station, Disposable Funnel 3521301 Mfr # G6001
COMPARE
47 250 Oil and Grease Extraction Individual Station, Disposable Funnel
₹251,000.00INR / Each
COMPARE
47 1000 Oil and Grease Extraction Individual Station, Glass Funnel; 47 mm
₹297,000.00INR / Each
COMPARE
90 1000 Oil and Grease Extraction Individual Station, Glass Funnel; 90 mm
₹344,000.00INR / Each