Environmental Express® SPE-Express® 2 Automated Oil & Grease System Accessories
Choose from a variety of additional accessories to expand and customize your system
  • Can string up to 3 stations together and operate simultaneously
The SPE-Express® 2 system is intended for the automated extraction of oil and grease material following US EPA Method 1664. The self-contained unit employs a Solid Phase Extraction (SPE) disk to capture and separate oil and grease components from sample media. Components are eluted from the SPE disk using n-hexane and collected in an aluminum pan. Following elution and collection, the remaining solvent mixture is then evaporated through gently applied heat source removing the n-hexane and capturing the oil and grease component residue.
360.00 - ₹69,500.00INR / Pkg of 50
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Aluminum Weigh Dish, 105 mm ; 100/Pk
₹6,300.00INR / Pkg of 100
COMPARE
Bottle Adapter, 53 mm
₹31,700.00INR / Each
COMPARE
Glass Chamber
₹62,100.00INR / Each
COMPARE
PTFE Reagent Manifold
₹13,700.00INR / Each
COMPARE
Reagent Bottle Rack
₹59,300.00INR / Each
COMPARE
Reagent Tubing Kit
₹69,500.00INR / Each
COMPARE
GL38 Cap, 4-Port PTFE Insert
₹5,700.00INR / Each
COMPARE
PTFE Port Plug, 5/16"-24 UNF(M)
₹2,000.00INR / Each
COMPARE
Hose Barb, Wye Reducer, 3/8" x 1/4" Inner Diameter
₹870.00INR / Each
COMPARE
Quick Disconnect Hose Barb Adapter
₹2,800.00INR / Each
COMPARE
3-Way Air Manifold
₹38,400.00INR / Each
COMPARE
47 mm Rinse Head Sub-Assembly
₹65,500.00INR / Each
COMPARE
90 mm Rinse Head Sub-Assembly
₹50,600.00INR / Each
COMPARE
Bottle Adapter for APC1430
₹35,700.00INR / Each
COMPARE
Bottle Adapter for APC1500
₹30,800.00INR / Each
COMPARE
Bottle Adapter, 70 mm
₹38,500.00INR / Each
COMPARE
Bottle Adapter, 89 mm
₹51,300.00INR / Each
COMPARE
UltraFlow® Filter Assembly, 47 mm, 100/Pack
₹45,400.00INR / Pkg of 100
COMPARE
UltraFlow® Filter Assembly, 90 mm, 50/Pack
₹67,500.00INR / Pkg of 50
COMPARE
5.5 g NaSO4 Drying Cartridge; 50/Pk
₹16,300.00INR / Pkg of 50
Showing 1 - 20 of 24