Environmental Express SPE-Express® 2 Hose Barb, Wye Reducer, 3/8" x 1/4" Inner Diameter
Representative image only.
Environmental Express SPE-Express® 2 Hose Barb, Wye Reducer, 3/8" x 1/4" Inner Diameter
Environmental Express | Item: 3539007
Choose from a variety of additional accessories to expand and customize your system
₹870.00INR / Each
₹870.00INR / Each
Product Description
Key Features
  • Can string up to 3 stations together and operate simultaneously
More About this Item
The SPE-Express® 2 system is intended for the automated extraction of oil and grease material following US EPA Method 1664. The self-contained unit employs a Solid Phase Extraction (SPE) disk to capture and separate oil and grease components from sample media. Components are eluted from the SPE disk using n-hexane and collected in an aluminum pan. Following elution and collection, the remaining solvent mixture is then evaporated through gently applied heat source removing the n-hexane and capturing the oil and grease component residue.
Specifications
  • DescriptionHose Barb, Wye Reducer, 3/8" x 1/4" Inner Diameter