Environmental Express® Plastic Ion Exchange Columns
Reduce sample contamination using these disposable columns
  • Perform ion exchanges with these unpacked disposable chromatography columns
  • Built-in shoulder provides greater surface area than standard columns
The clarified polypropylene columns have a built-in polyethylene frit with nominal 30 µm particle retention. The unique design sets the frit slightly above the bottom, creating a “shoulder” that increases surface area for more efficient resin separations.
Order funnels separately for ease of fill—funnels attach directly to column entrance.
9,800.00 - ₹28,100.00INR / Pkg of 50
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (mL) End Cap Fritted Disk
Availability Pricing
COMPARE
2.5 No Polyethylene, 50 µm retention size
₹12,700.00INR / Pkg of 50
COMPARE
2.5 Yes Polyethylene, 30 µm retention size
₹9,800.00INR / Pkg of 50
COMPARE
5.5 No Polyethylene, 50 µm retention size
₹19,500.00INR / Pkg of 50
COMPARE
5.5 Yes Polyethylene, 50 µm retention size
₹16,600.00INR / Pkg of 50
COMPARE
18.5 Yes Polyethylene, 50 µm retention size
₹28,100.00INR / Pkg of 50