Environmental Express® Oil and Grease Extraction Disks
Compliant with EPA method 1664 for SPE of oil and grease
  • Varied materials to choose from
  • Diameter sizes for any application
Choice of nylon, glass microfiber, UltraFlow C18, or preactivated UltraPrep.
5,600.00 - ₹39,200.00INR / Pkg of 20
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material
Availability Pricing
COMPARE
47 Glass Microfiber
₹10,100.00INR / Pkg of 50
COMPARE
90 Glass Microfiber
₹13,300.00INR / Pkg of 50
COMPARE
47 UltraFlow C18
₹21,800.00INR / Pkg of 20
COMPARE
47 Nylon
₹5,600.00INR / Pkg
COMPARE
90 UltraFlow C18
₹29,900.00INR / Pkg of 15
COMPARE
100 UltraFlow C18
₹32,900.00INR / Pkg of 15
COMPARE
100 UltraLow C18
₹29,600.00INR / Pkg of 15
COMPARE
47 UltraLow C18
₹19,500.00INR / Pkg of 20
COMPARE
90 UltraLow C18
₹27,300.00INR / Pkg of 15
COMPARE
47 UltraPrep (preactivated)
₹26,000.00INR / Pkg of 20
COMPARE
90 UltraPrep (preactivated)
₹35,100.00INR / Pkg of 15
COMPARE
100 UltraPrep (preactivated)
₹39,200.00INR / Pkg of 15
Showing 1- 12 of 12