Environmental Express® IAQ 15 Connect™ Portable Air Sampling Pump Kits
An advanced portable, battery-powered air sampling pump designed for exclusive use with Air-O-Cell® and Via-Cell® cassettes for fast sample collection
  • Compact design allows access to crawl spaces, HVAC vents, and other confined sampling areas
  • Long-lasting battery for up to 99 five-minute samples
  • The only sampling pump with IAQ specific calibration verification and accuracy at 15 L/min
  • LED screen with button operated navigation and on-demand access to all sampling controls for ease of use
  • Various sampling durations (1, 2, 5, or 10 minutes) for any sampling setting
  • Automatic event logging keeps track of critical sampling data with automatic data logging and Bluetooth® connection IAQ sampling app control
  • Retrieve sampling records with the IAQ 15 app, available on the Apple App Store and Google Play
The IAQ 15 Connect is a portable air sampling pump meant for use with Air-O-Cell® and Via-Cell® cassettes to make doing your job easier. The pump has three sampling modes: quick sample, single sampling, and sequential sampling. Quick sample mode allows easy sampling with the most used sampling times of 1, 2, 5, or 10 minutes. Single sampling mode is designed to take one sample at a predetermined amount of time. The sampling time can be set anywhere from 1 minute to 9 hours, 59 minutes. Sequential sampling mode allows the pump to be programmed to turn on and off at predetermined times. The user programs the number of on/off cycles, the cycle run time, and the cycle off time.
Calibration is made easy with the supplied flow indicator for Air-O-Cell cassettes. You may also calibrate the pump using an optional TSI® 4046 Primary Calibrator with attachments or using an optional Via-Cell flow indicator (if sampling with Via-Cell cassettes).
The IAQ 15 Connect pump also pairs with the IAQ 15 app, available on the Apple App Store and Google Play. Through Bluetooth® connection, the app can retrieve multiple records from the pump, including location, sampling type, cycle count, run-time, calibration status, RFID information, and more.
126,000.00 - ₹132,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Portable Air Sampling Pump Kit
₹126,000.00INR / Each
COMPARE
Pro Portable Air Sampling Pump Kit
₹132,000.00INR / Each