Environmental Express® Glass Fiber Filters
Use for air and water pollution official test methods
  • High loading capacity
  • Withstands high temperatures up to 550°C
  • Collection equivalent to Pall® Gelman A/E, Whatman™ 934-AH™, and Millipore® AP40 filters
  • Typically used for testing such as TSS, TDS, and VSS in wastewater, as outlined in Standard Methods 2540*
Filters are made from unprepared, binderless, borosilicate glass and have a 1.5 µm pore size — ideal for water and air pollution testing methods, scintillation counting, and cell harvesting. Each filter is laser cut to ensure a clean, sealed, fiber-free edge that maintains its integrity throughout your entire testing process. Conveniently packaged in a snap-cap tube of 100 filters.
Autoclavable
*Filters are suitable for Standard Methods 2540 but will require preparation before use.
4,900.00 - ₹32,800.00INR / Pkg of 100
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Pore Size (micron)
Availability Pricing
COMPARE
37 1.5
₹7,800.00INR / Pkg of 100
COMPARE
21 1.5
₹5,600.00INR / Pkg of 100
COMPARE
24 1.5
₹6,100.00INR / Pkg of 100
COMPARE
25 1.5
₹6,500.00INR / Pkg of 100
COMPARE
32 1.5
₹7,000.00INR / Pkg of 100
COMPARE
35 1.5
₹7,700.00INR / Pkg of 100
COMPARE
43 1.5
₹8,900.00INR / Pkg of 100
COMPARE
47 1.5
₹4,900.00INR / Pkg of 100
COMPARE
55 1.5
₹9,700.00INR / Pkg of 100
COMPARE
70 1.5
₹12,800.00INR / Pkg of 100
COMPARE
90 1.5
₹13,200.00INR / Pkg of 100
COMPARE
102 1.5
₹16,700.00INR / Pkg of 100
COMPARE
110 1.5
₹21,400.00INR / Pkg of 100
COMPARE
125 1.5
₹23,400.00INR / Pkg of 100
COMPARE
142 1.5
₹31,500.00INR / Pkg of 100
COMPARE
150 1.5
₹32,800.00INR / Pkg of 100
Showing 1- 16 of 16