Environmental Express® Dairy-Style Jugs, HDPE
Grab and pour with easy-grip handle
  • Leakproof design keeps contents safe inside
  • Familiar and easy pouring with the milk-type jug design
  • Pre-cleaned for immediate use
Included caps are composed of polypropylene (PP) and lined with F217, a low-density foam core between two solid layers of LDPE. Level 3 pre-cleaned containers meet EPA guidelines, but do not ship with a Certificate of Analysis.
7,900.00 - ₹38,500.00INR / Case of 108
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Material
Availability Pricing
COMPARE
1 38 mm High Density Polyethyene (HDPE)
₹38,500.00INR / Case of 108
COMPARE
2 38 mm High Density Polyethyene (HDPE)
₹15,100.00INR / Case of 40
COMPARE
3.845 38 mm High Density Polyethyene (HDPE)
₹7,900.00INR / Case of 20