Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene
Threaded barb adapter design provides safe, difficult-to-break connection
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Single barb design tightly “grips” the tubing
  • RoHS compliant
Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. The elbows feature a 90° bend and is designed to fit in a tight space. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place.
These animal derivative-free polypropylene fittings are radiation stable and resistant to solvents, chemicals, water, strong mineral acids, bases, and other inorganic environments. Not recommended for use with oxidizing agents.
1,600.00 - ₹5,100.00INR / Pkg of 10
46 variations of this product are available.
See All Product Options Below
46 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Flow Pattern Connection Type
Availability Pricing
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 3/4" NPT(M) x 5/8" ID; 10/PK 5012681 Mfr # L12-10PP
COMPARE
5/8 90° elbow Threaded NPT(M) x Hose Barb
₹2,500.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 3/4" NPT(M) x 3/4" ID; 10/PK 5012682 Mfr # L12-12PP
COMPARE
3/4 90° elbow Threaded NPT(M) x Hose Barb
₹2,000.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 3/4" NPT(M) x 1" ID; 10/PK 5012683 Mfr # L12-16PP
COMPARE
1 90° elbow Threaded NPT(M) x Hose Barb
₹2,500.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 3/4" NPT(M) x 1/4" ID; 10/PK 5012685 Mfr # L12-4PP
COMPARE
1/4 90° elbow Threaded NPT(M) x Hose Barb
₹2,000.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 3/4" NPT(M) x 3/8" ID; 10/PK 5012686 Mfr # L12-6PP
COMPARE
3/8 90° elbow Threaded NPT(M) x Hose Barb
₹2,400.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 3/4" NPT(M) x 1/2" ID; 10/PK 5012687 Mfr # L12-8PP
COMPARE
1/2 90° elbow Threaded NPT(M) x Hose Barb
₹2,400.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 1/4"-28 UNF x 1/8" ID; 10/PK 5012688 Mfr # L1428-2-200PP
COMPARE
1/8 90° elbow Threaded UNF x Hose Barb
₹2,300.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®  1/8" NPT x 3/32" ID; 10/PK 5012689 Mfr # L2-1.5-200NK7
COMPARE
3/32 90° elbow Threaded NPT(M) x Hose Barb
₹5,100.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 1/4"-28 UNF x 3/16" ID; 10/PK 5012690 Mfr # L1428-3-200PP
COMPARE
3/16 90° elbow Threaded UNF x Hose Barb
₹3,400.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 1" NPT(M) x 3/4" ID; 10/PK 5012691 Mfr # L16-12PP
COMPARE
3/4 90° elbow Threaded NPT(M) x Hose Barb
₹2,800.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 1" NPT(M) x 1" ID; 10/PK 5012692 Mfr # L16-16PP
COMPARE
1 90° elbow Threaded NPT(M) x Hose Barb
₹2,800.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 1/8" NPT(M) x 3/32" ID; 10/PK 5012693 Mfr # L2-1.5-200PP
COMPARE
3/32 90° elbow Threaded NPT(M) x Hose Barb
₹3,300.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 1/8" NPT(M) x 1/16" ID; 10/PK 5012694 Mfr # L2-1-200PP
COMPARE
1/16 90° elbow Threaded NPT(M) x Hose Barb
₹3,300.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 1/8" NPT(M) x 5/32" ID; 10/PK 5012695 Mfr # L2-2.5PP
COMPARE
5/32 90° elbow Threaded NPT(M) x Hose Barb
₹1,800.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 1/8" NPT(M) x 1/8" ID; 10/PK 5012696 Mfr # L2-2PP
COMPARE
1/8 90° elbow Threaded NPT(M) x Hose Barb
₹1,800.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 1/8" NPT(M) x 3/16" ID; 10/PK 5012697 Mfr # L2-3PP
COMPARE
3/16 90° elbow Threaded NPT(M) x Hose Barb
₹1,800.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 1/8" NPT(M) x 1/4" ID; 10/PK 5012698 Mfr # L2-4PP
COMPARE
1/4 90° elbow Threaded NPT(M) x Hose Barb
₹1,800.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 1/8" NPT(M) x 5/16" ID; 10/PK 5012699 Mfr # L2-5PP
COMPARE
5/16 90° elbow Threaded NPT(M) x Hose Barb
₹1,800.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 1/8" NPT(M) x 3/8" ID; 10/PK 5012700 Mfr # L2-6PP
COMPARE
3/8 90° elbow Threaded NPT(M) x Hose Barb
₹1,800.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 1/8" BSPT(M) x 1/8" ID; 10/PK 5012701 Mfr # L2B-2PP
COMPARE
1/8 90° elbow Threaded BSPT(M) x Hose Barb
₹1,600.00INR / Pkg of 10
Showing 1 - 20 of 46