Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 3/4" NPT(M) x 5/8" ID; 10/PK
Representative image only.
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 3/4" NPT(M) x 5/8" ID; 10/PK
Eldon James | Mfr: L12-10PP| Item: 5012681
Threaded barb adapter design provides safe, difficult-to-break connection
₹2,500.00INR / Pkg of 10
₹2,500.00INR / Pkg of 10
Buy It Together
Current Item
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 3/4" NPT(M) x 5/8" ID; 10/PK
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 3/4" NPT(M) x 5/8" ID; 10/PK
5012681
₹2,500.00INR / Pkg of 10
+ + + =
Total Price 11100.00 ADD 4 ITEMS TO CART
Product Description
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Single barb design tightly “grips” the tubing
  • RoHS compliant
  • 3/4" NPT(M) x 5/8" hose barb
More About this Item
Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. The elbows feature a 90° bend and is designed to fit in a tight space. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place.
These animal derivative-free polypropylene fittings are radiation stable and resistant to solvents, chemicals, water, strong mineral acids, bases, and other inorganic environments. Not recommended for use with oxidizing agents.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPolypropylene
  • Sterilization MethodGamma irradiation between 25 and 50 kGy, EtO, or autoclave up to 121°C (250°F)
  • Connection TypeThreaded NPT(M) x Hose Barb
  • Number Of Ports2
  • Thread Size NPT(M) (Port 1) (InchShort)3/4
  • Barb Size / Tubing Id (Port 2) (in)5/8
  • Flow Pattern90° elbow
  • ColorNatural
  • Min Temperature (° F)-9
  • Max Temperature (° F)150
  • Min Temperature (° C)-23
  • Max Temperature (° C)66
  • Regulatory ApprovalsADCF;FDA;Non-Phthalate;Non-DEHP;RoHS
  • DescriptionThreaded to Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene, 3/4" NPT(M) x 5/8" ID; 10/PK