Eldon James Threaded NPT(M) to Hose Barb Fittings, Straight Adapter, Stainless Steel
Threaded barb adapter design provides safe, difficult-to-break connection
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Single barb design tightly “grips” the tubing
  • RoHS compliant
Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place.
These durable stainless steel fittings perform well in high temperatures and challenging environments. Made of 316L molybdenum bearing stainless steel, they are widely used in pharmaceutical, food, and beverage applications. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
3,300.00 - ₹5,600.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Fitting Material Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Stainless Steel, 1/8" NPT(M) x 1/8" ID; 1/Each 5012459 Mfr # A2-2SS
COMPARE
1/8 Stainless Steel 1/8
₹5,600.00INR / Each
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Stainless Steel, 1/8" NPT(M) x 1/4" ID; 1/Each 5012460 Mfr # A2-4SS
COMPARE
1/4 Stainless Steel 1/8
₹3,300.00INR / Each
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Stainless Steel, 1/8" NPT(M) x 5/16" ID; 1/Each 5012461 Mfr # A2-5SS
COMPARE
5/16 Stainless Steel 1/8
₹3,500.00INR / Each
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Stainless Steel, 1/4" NPT(M) x 1/4" ID; 1/Each 5012462 Mfr # A4-4SS
COMPARE
1/4 Stainless Steel 1/4
₹3,400.00INR / Each
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Stainless Steel, 1/4" NPT(M) x 5/16" ID; 1/Each 5012463 Mfr # A4-5SS
COMPARE
5/16 Stainless Steel 1/4
₹3,600.00INR / Each