Eldon James Threaded NPT(M) to Hose Barb Fittings, Straight Adapter, Natural Nylon
Threaded barb adapter design provides safe, difficult-to-break connection
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Single barb design tightly “grips” the tubing
  • RoHS compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place. An anti-rotation device on the hose barb prevents tubing from rotating, minimizing kinks.
These natural nylon fittings provide good tensile strength, stiffness, and impact resistance and are resistant to fuels
1,500.00 - ₹2,700.00INR / Pkg of 10
30 variations of this product are available.
See All Product Options Below
30 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Flow Pattern Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 3/4" NPT(M) x 5/8" ID; 10/PK 5012399 Mfr # A12-10NN
COMPARE
5/8 Straight 3/4
₹1,800.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 3/4" NPT(M) x 3/4" ID; 10/PK 5012400 Mfr # A12-12NN
COMPARE
3/4 Straight 3/4
₹2,500.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 3/4" NPT(M) x 1" ID; 10/PK 5012401 Mfr # A12-16NN
COMPARE
1 Straight 3/4
₹2,700.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 3/4" NPT(M) x 1/4" ID; 10/PK 5012402 Mfr # A12-4NN
COMPARE
1/4 Straight 3/4
₹1,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 3/4" NPT(M) x 3/8" ID; 10/PK 5012403 Mfr # A12-6NN
COMPARE
3/8 Straight 3/4
₹1,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 3/4" NPT(M) x 1/2" ID; 10/PK 5012404 Mfr # A12-8NN
COMPARE
1/2 Straight 3/4
₹1,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 1" NPT(M) x 1" ID; 10/PK 5012405 Mfr # A16-16NN
COMPARE
1 Straight 1
₹2,700.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 1/8" NPT(M) x 5/32" ID; 10/PK 5012406 Mfr # A2-2.5NN
COMPARE
5/32 Straight 1/8
₹1,500.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 1/8" NPT(M) x 1/8" ID; 10/PK 5012407 Mfr # A2-2NN
COMPARE
1/8 Straight 1/8
₹1,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 1/8" NPT(M) x 3/16" ID; 10/PK 5012408 Mfr # A2-3NN
COMPARE
3/16 Straight 1/8
₹1,500.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 1/8" NPT(M) x 1/4" ID; 10/PK 5012409 Mfr # A2-4NN
COMPARE
1/4 Straight 1/8
₹1,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 1/8" NPT(M) x 5/16" ID; 10/PK 5012410 Mfr # A2-5NN
COMPARE
5/16 Straight 1/8
₹1,500.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 1/8" NPT(M) x 3/8" ID; 10/PK 5012411 Mfr # A2-6NN
COMPARE
3/8 Straight 1/8
₹1,700.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 1/4" NPT(M) x 5/32" ID; 10/PK 5012412 Mfr # A4-2.5NN
COMPARE
5/32 Straight 1/4
₹1,500.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 1/4" NPT(M) x 1/8" ID; 10/PK 5012413 Mfr # A4-2NN
COMPARE
1/8 Straight 1/4
₹1,800.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 1/4" NPT(M) x 3/16" ID; 10/PK 5012414 Mfr # A4-3NN
COMPARE
3/16 Straight 1/4
₹1,500.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 1/4" NPT(M) x 1/4" ID; 10/PK 5012415 Mfr # A4-4NN
COMPARE
1/4 Straight 1/4
₹1,500.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 1/4" NPT(M) x 5/16" ID; 10/PK 5012416 Mfr # A4-5NN
COMPARE
5/16 Straight 1/4
₹1,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 1/4" NPT(M) x 3/8" ID; 10/PK 5012417 Mfr # A4-6NN
COMPARE
3/8 Straight 1/4
₹1,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 1/4" NPT(M) x 1/2" ID; 10/PK 5012418 Mfr # A4-8NN
COMPARE
1/2 Straight 1/4
₹1,600.00INR / Pkg of 10
Showing 1 - 20 of 30